Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025408.2 | NP_079684.2 | 4 - 5 | 398 | 176 | |
GenBank mRNA | AK011668.1 | - | 4 - 5 | 379 | 176 | |
AK017361.1 | - | 303 | 176 | |||
AK080977.1 | - | 4 - 5 | 376 | 176 | ||
AK150629.1 | - | 4 - 5 | 359 | 176 | ||
AK162403.1 | - | 4 - 5 | 414 | 176 | ||
BC023924.1 | - | 4 - 5 | 402 | 5324078 | 176 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |