Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005013.2 | - | 1 - 2 | 285 | 84 | |
AK017378.1 | - | 1 - 2 | 292 | 84 | ||
BC006613.1 | - | 1 - 2 | 279 | 3967332 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |