Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009099.2 | NP_033125.2 | 1 - 2 | 187 | 89 | |
GenBank mRNA | AK089726.1 | - | 1 - 2 | 184 | 89 | |
AK134144.1 | - | 1 - 2 | 156 | 89 | ||
AK137506.1 | - | 1 - 2 | 188 | 89 | ||
AK138249.1 | - | 1 - 2 | 190 | 89 | ||
BC005447.1 | - | 1 - 2 | 121 | 3493256 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |