Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161724.1 | NP_001155196.1 | 1 - 2 | 128 | 82 | |
GenBank mRNA | AK002344.1 | - | 1 - 2 | 46 | 82 | |
AK011766.1 | - | 1 - 2 | 66 | 82 | ||
AK154207.1 | - | 1 - 2 | 49 | 82 | ||
CA752166.1 | - | 1 - 2 | 128 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |