Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172908.3 | NP_766496.2 | 2 - 3 | 197 | 76 | |
GenBank mRNA | AK033776.1 | - | 1 - 2 | 86 | 76 | |
BC125284.1 | - | 2 - 3 | 226 | 40129799 | 76 | |
BC125288.1 | - | 2 - 3 | 226 | 40129803 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |