Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172908.3 | NP_766496.2 | 12 - 13 | 1414 | 110 | |
GenBank mRNA | AK033776.1 | - | 11 - 12 | 1303 | 110 | |
BC125284.1 | - | 12 - 13 | 1443 | 40129799 | 110 | |
BC125288.1 | - | 12 - 13 | 1443 | 40129803 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |