Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025344.2 | NP_079620.2 | 3 - 4 | 546 | 114 | |
GenBank mRNA | AK002778.1 | - | 3 - 4 | 545 | 114 | |
AK003372.1 | - | 3 - 4 | 544 | 114 | ||
AK010333.1 | - | 3 - 4 | 540 | 114 | ||
AK088537.1 | - | 3 - 4 | 546 | 114 | ||
AK136797.1 | - | 545 | 114 | |||
AK152037.1 | - | 3 - 4 | 547 | 114 | ||
AK190701.1 | - | 3 - 4 | 191 | 114 | ||
AK197956.1 | - | 3 - 4 | 193 | 114 | ||
AK217928.1 | - | 2 - 3 | 164 | 114 | ||
BC070473.1 | - | 3 - 4 | 528 | 30634857 | 114 | |
BC083190.1 | - | 3 - 4 | 531 | 30358339 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |