Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025559.3 | NP_001020730.1 | 3 - 4 | 375 | 94 | |
NM_001025560.2 | NP_001020731.1 | 3 - 4 | 495 | 94 | ||
NM_001277326.1 | NP_001264255.1 | 2 - 3 | 350 | 94 | ||
NM_001277327.1 | NP_001264256.1 | 2 - 3 | 530 | 94 | ||
NM_001277328.1 | NP_001264257.1 | 1 - 2 | 238 | 94 | ||
NM_011445.4 | NP_035575.1 | 3 - 4 | 403 | 94 | ||
GenBank mRNA | AK029696.1 | - | 1 - 2 | 165 | 94 | |
AK030713.1 | - | 3 - 4 | 496 | 94 | ||
AK048143.1 | - | 3 - 4 | 376 | 94 | ||
AK084290.1 | - | 3 - 4 | 534 | 94 | ||
AK132409.1 | - | 2 - 3 | 316 | 94 | ||
BC067407.1 | - | 2 - 3 | 527 | 30094413 | 94 | |
D61689.1 | - | 1 - 2 | 234 | 94 | ||
U32614.1 | - | 3 - 4 | 407 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |