Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146198.2 | NP_666310.1 | 9 - 10 | 937 | 81 | |
GenBank mRNA | AF518726.1 | - | 10 - 11 | 1030 | 81 | |
AK008219.1 | - | 9 - 10 | 991 | 81 | ||
BC031742.1 | - | 9 - 10 | 938 | 4221537 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |