Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301374.1 | NP_001288303.1 | 2 - 3 | 457 | 100 | |
NM_009898.3 | NP_034028.1 | 2 - 3 | 370 | 100 | ||
GenBank mRNA | AF047388.1 | - | 1 - 2 | 204 | 100 | |
AF143955.1 | - | 2 - 3 | 341 | 100 | ||
AF495468.1 | - | 2 - 3 | 308 | 100 | ||
AK040885.1 | - | 2 - 3 | 313 | 100 | ||
AK075799.1 | - | 2 - 3 | 305 | 100 | ||
AK146290.1 | - | 2 - 3 | 308 | 100 | ||
AK150962.1 | - | 2 - 3 | 312 | 100 | ||
AK151755.1 | - | 2 - 3 | 312 | 100 | ||
AK155527.1 | - | 2 - 3 | 313 | 100 | ||
AK155860.1 | - | 2 - 3 | 308 | 100 | ||
AK165296.1 | - | 2 - 3 | 312 | 100 | ||
AK172447.1 | - | 2 - 3 | 308 | 100 | ||
BC002136.1 | - | 2 - 3 | 298 | 3484185 | 100 | |
BC053398.1 | - | 2 - 3 | 288 | 30076939 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |