Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008136.2 | NP_032162.2 | 2 - 3 | 457 | 84 | |
GenBank mRNA | AK078187.1 | - | 2 - 3 | 84 | ||
BC010298.1 | - | 2 - 3 | 412 | 3155411 | 84 | |
BC013243.1 | - | 2 - 3 | 481 | 84 | ||
BM949933.1 | - | 2 - 3 | 203 | 84 | ||
X65026.1 | - | 1 - 2 | 100 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |