Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183168.2 | NP_898991.1 | 1 - 2 | 149 | 61 | |
GenBank mRNA | AK142680.1 | - | 1 - 2 | 174 | 61 | |
AX775901.1 | - | 1 - 2 | 109 | 61 | ||
BC027331.1 | - | 1 - 2 | 145 | 3982042 | 61 | |
BC064095.1 | - | 1 - 2 | 129 | 1054566 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |