Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008027.2 | NP_032053.1 | 7 - 8 | 726 | 73 | |
GenBank mRNA | AF145044.1 | - | 6 - 7 | 567 | 73 | |
AK150692.1 | - | 7 - 8 | 748 | 73 | ||
AK150827.1 | - | 7 - 8 | 675 | 73 | ||
AK167323.1 | - | 7 - 8 | 653 | 73 | ||
AY167925.1 | - | 7 - 8 | 732 | 73 | ||
BC004647.1 | - | 7 - 8 | 736 | 3158071 | 73 | |
U90435.1 | - | 7 - 8 | 726 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |