Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tial1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009383.2 | NP_033409.1 | 8 - 9 | 1060 | 116 | |
GenBank mRNA | AK012085.1 | - | 8 - 9 | 704 | 116 | |
AK054473.1 | - | 2 - 3 | 122 | 116 | ||
AK216078.1 | - | 2 - 3 | 130 | 116 | ||
BC010496.1 | - | 9 - 10 | 794 | 3497971 | 116 | |
BC068194.1 | - | 4 - 5 | 336 | 116 | ||
U55861.1 | - | 8 - 9 | 836 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |