Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114322.1 | NP_001107794.1 | 6 - 7 | 698 | 108 | |
NM_028069.3 | NP_082345.1 | 6 - 7 | 698 | 108 | ||
GenBank mRNA | AF462391.1 | - | 6 - 7 | 687 | 108 | |
AK007988.1 | - | 6 - 7 | 687 | 108 | ||
AK027913.1 | - | 6 - 7 | 699 | 108 | ||
BC043336.1 | - | 6 - 7 | 663 | 5042289 | 108 | |
BC054469.1 | - | 6 - 7 | 683 | 4225164 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |