Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114322.1 | NP_001107794.1 | 11 - 12 | 1370 | 72 | |
NM_028069.3 | NP_082345.1 | 11 - 12 | 1370 | 72 | ||
GenBank mRNA | AF462391.1 | - | 11 - 12 | 1359 | 72 | |
AK007988.1 | - | 11 - 12 | 1359 | 72 | ||
AK027913.1 | - | 11 - 12 | 1371 | 72 | ||
BC043336.1 | - | 11 - 12 | 1335 | 5042289 | 72 | |
BC054469.1 | - | 11 - 12 | 1355 | 4225164 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |