Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025826.4 | NP_080102.1 | 1 - 2 | 184 | 81 | |
GenBank mRNA | AK004889.1 | - | 1 - 2 | 85 | 81 | |
AK076535.1 | - | 1 - 2 | 85 | 81 | ||
AK131628.1 | - | 1 - 2 | 131 | 81 | ||
AK152325.1 | - | 1 - 2 | 75 | 81 | ||
BC011235.1 | - | 1 - 2 | 65 | 81 | ||
BC044826.1 | - | 1 - 2 | 118 | 81 | ||
BC054428.1 | - | 1 - 2 | 126 | 5039627 | 81 | |
BY296718.1 | - | 1 - 2 | 185 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |