Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028844.3 | NP_083120.2 | 1 - 2 | 259 | 73 | |
GenBank mRNA | AK006808.1 | - | 1 - 2 | 260 | 73 | |
BC100735.1 | - | 2 - 3 | 136 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons |