Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145820.1 | NP_001139292.1 | 6 - 7 | 986 | 66 | |
NM_010274.3 | NP_034404.3 | 6 - 7 | 861 | 66 | ||
GenBank mRNA | AK034353.1 | - | 6 - 7 | 770 | 66 | |
AK144716.1 | - | 6 - 7 | 987 | 66 | ||
AK151189.1 | - | 6 - 7 | 773 | 66 | ||
AK167152.1 | - | 6 - 7 | 799 | 66 | ||
BC021359.1 | - | 6 - 7 | 955 | 5012142 | 66 | |
D50430.1 | - | 6 - 7 | 763 | 66 | ||
U60987.1 | - | 6 - 7 | 797 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |