Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252572.1 | NP_001239501.1 | 15 - 16 | 1660 | 80 | |
NM_007803.5 | NP_031829.2 | 16 - 17 | 1771 | 80 | ||
GenBank mRNA | AK084249.1 | - | 15 - 16 | 1470 | 80 | |
AK146856.1 | - | 16 - 17 | 1578 | 80 | ||
AK148010.1 | - | 16 - 17 | 1574 | 80 | ||
AK165555.1 | - | 15 - 16 | 1463 | 80 | ||
AK168699.1 | - | 16 - 17 | 1572 | 80 | ||
AK168871.1 | - | 15 - 16 | 1461 | 80 | ||
AK168945.1 | - | 15 - 16 | 1471 | 80 | ||
AK185163.1 | - | 3 - 4 | 80 | |||
BC011434.1 | - | 15 - 16 | 1453 | 3981559 | 80 | |
BC117820.1 | - | 15 - 16 | 1473 | 80 | ||
U03184.1 | - | 16 - 17 | 1560 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |