Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252572.1 | NP_001239501.1 | 1 - 2 | 246 | 70 | |
NM_007803.5 | NP_031829.2 | 1 - 2 | 246 | 70 | ||
GenBank mRNA | AK084249.1 | - | 1 - 2 | 56 | 70 | |
AK146856.1 | - | 1 - 2 | 53 | 70 | ||
AK148010.1 | - | 1 - 2 | 49 | 70 | ||
AK165555.1 | - | 1 - 2 | 49 | 70 | ||
AK168699.1 | - | 1 - 2 | 47 | 70 | ||
AK168871.1 | - | 1 - 2 | 47 | 70 | ||
AK168945.1 | - | 1 - 2 | 57 | 70 | ||
BC011434.1 | - | 1 - 2 | 39 | 3981559 | 70 | |
BY137811.1 | - | 1 - 2 | 246 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |