Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111049.1 | NP_001104519.1 | 2 - 3 | 237 | 67 | |
NM_009842.3 | NP_033972.2 | 2 - 3 | 355 | 67 | ||
GenBank mRNA | BQ896459.1 | - | 2 - 3 | 373 | 67 | |
DV063036.1 | - | 2 - 3 | 264 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |