Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146206.4 | NP_666318.2 | 11 - 12 | 1029 | 57 | |
GenBank mRNA | AK040854.1 | - | 11 - 12 | 1057 | 57 | |
AK052930.1 | - | 11 - 12 | 1030 | 57 | ||
AK089490.1 | - | 11 - 12 | 1024 | 57 | ||
AK090059.1 | - | 11 - 12 | 1033 | 57 | ||
AK170863.1 | - | 11 - 12 | 1025 | 57 | ||
BC141195.1 | - | 11 - 12 | 1113 | 9055807 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |