Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127363.1 | NP_001120835.1 | 8 - 9 | 921 | 100 | |
NM_183144.3 | NP_898967.2 | 8 - 9 | 921 | 100 | ||
GenBank mRNA | AK083449.1 | - | 9 - 10 | 920 | 100 | |
AK157558.1 | - | 8 - 9 | 784 | 100 | ||
AK171360.1 | - | 8 - 9 | 921 | 100 | ||
AK171880.1 | - | 8 - 9 | 897 | 100 | ||
BC056341.1 | - | 8 - 9 | 921 | 5703345 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |