Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003915.2 | NP_001003915.1 | 6 - 7 | 1127 | 100 | |
NM_001177624.1 | NP_001171095.1 | 6 - 7 | 1127 | 100 | ||
GenBank mRNA | AF529222.1 | - | 6 - 7 | 1135 | 100 | |
AK085356.1 | - | 6 - 7 | 1120 | 100 | ||
AK128951.1 | - | 6 - 7 | 1119 | 100 | ||
AK144004.1 | - | 6 - 7 | 1060 | 100 | ||
AK165419.1 | - | 6 - 7 | 1120 | 100 | ||
AY964639.1 | - | 6 - 7 | 830 | 100 | ||
BC121791.1 | - | 6 - 7 | 823 | 100 | ||
BC141255.1 | - | 6 - 7 | 1055 | 9055867 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |