Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033319.2 | NP_001028491.1 | 29 - 30 | 3877 | 75 | |
NM_001195086.1 | NP_001182015.1 | 30 - 31 | 3952 | 75 | ||
GenBank mRNA | AK160345.1 | - | 2 - 3 | 213 | 75 | |
BC038349.1 | - | 16 - 17 | 2150 | 75 | ||
BC157936.1 | - | 30 - 31 | 3769 | 9088116 | 75 | |
BC158007.1 | - | 29 - 30 | 3834 | 9007315 | 75 | |
BC172096.1 | - | 30 - 31 | 3772 | 9054382 | 75 | |
EU568869.1 | - | 30 - 31 | 3955 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |