Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033319.2 | NP_001028491.1 | 10 - 11 | 1505 | 79 | |
NM_001195086.1 | NP_001182015.1 | 11 - 12 | 1580 | 79 | ||
GenBank mRNA | BC157936.1 | - | 11 - 12 | 1397 | 9088116 | 79 |
BC158007.1 | - | 10 - 11 | 1462 | 9007315 | 79 | |
BC172096.1 | - | 11 - 12 | 1400 | 9054382 | 79 | |
EU568869.1 | - | 11 - 12 | 1580 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |