Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178755.3 | NP_848870.2 | 5 - 6 | 437 | 60 | |
GenBank mRNA | AK029744.1 | - | 5 - 6 | 470 | 60 | |
AK029932.1 | - | 5 - 6 | 438 | 60 | ||
AK030138.1 | - | 5 - 6 | 458 | 60 | ||
AK040259.1 | - | 5 - 6 | 373 | 60 | ||
AK079691.1 | - | 5 - 6 | 374 | 60 | ||
DQ867026.1 | - | 5 - 6 | 470 | 60 | ||
DQ867027.1 | - | 5 - 6 | 470 | 60 | ||
DQ867028.1 | - | 5 - 6 | 470 | 60 | ||
DQ867029.1 | - | 5 - 6 | 470 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |