Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290649.1 | NP_001277578.1 | 1 - 2 | 160 | 148 | |
NM_029793.2 | NP_084069.1 | 1 - 2 | 160 | 148 | ||
NR_110968.1 | 1 - 2 | 160 | 148 | |||
GenBank mRNA | AK002954.1 | - | 1 - 2 | 104 | 148 | |
AK028516.1 | - | 1 - 2 | 79 | 148 | ||
AK044987.1 | - | 1 - 2 | 108 | 148 | ||
BC024637.1 | - | 1 - 2 | 82 | 148 | ||
BC053068.1 | - | 1 - 2 | 88 | 6405595 | 148 | |
BC080733.1 | - | 1 - 2 | 160 | 30110235 | 148 | |
BY084848.1 | - | 1 - 2 | 155 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |