Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178778.3 | NP_848893.2 | 12 - 13 | 1226 | 121 | |
GenBank mRNA | AK044772.1 | - | 12 - 13 | 1222 | 121 | |
AK082408.1 | - | 13 - 14 | 1328 | 121 | ||
BC132485.1 | - | 12 - 13 | 1201 | 40130762 | 121 | |
BC144986.1 | - | 13 - 14 | 1302 | 121 | ||
BC144987.1 | - | 12 - 13 | 1277 | 121 | ||
BC144989.1 | - | 12 - 13 | 1200 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |