Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178890.3 | NP_849221.2 | 5 - 6 | 1979 | 89 | |
GenBank mRNA | AK042322.1 | - | 5 - 6 | 1978 | 90 | |
AK151620.1 | - | 5 - 6 | 1975 | 89 | ||
AK152163.1 | - | 5 - 6 | 1975 | 89 | ||
AK170569.1 | - | 5 - 6 | 1978 | 89 | ||
AK172414.1 | - | 5 - 6 | 1977 | 89 | ||
AK172468.1 | - | 5 - 6 | 1976 | 89 | ||
BC054399.1 | - | 5 - 6 | 1979 | 6493840 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |