Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173749.4 | NP_776110.3 | 6 - 7 | 992 | 69 | |
GenBank mRNA | AA444868.1 | - | 2 - 3 | 239 | 69 | |
AK031254.1 | - | 6 - 7 | 864 | 69 | ||
AK088017.1 | - | 6 - 7 | 891 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |