Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mep1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008585.2 | NP_032611.2 | 1 - 2 | 120 | 133 | |
GenBank mRNA | AK093914.1 | - | 1 - 2 | 81 | 133 | |
AK143906.1 | - | 1 - 2 | 77 | 133 | ||
AK164734.1 | - | 1 - 2 | 75 | 133 | ||
BC015258.1 | - | 1 - 2 | 120 | 4241252 | 133 | |
M74897.1 | - | 1 - 2 | 119 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |