Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028711.3 | NP_082987.1 | 2 - 3 | 315 | 72 | |
GenBank mRNA | AB106930.1 | - | 2 - 3 | 459 | 72 | |
AK014394.1 | - | 2 - 3 | 316 | 72 | ||
AK043831.1 | - | 2 - 3 | 515 | 72 | ||
AK054292.1 | - | 1 - 2 | 878 | 72 | ||
BC138993.1 | - | 2 - 3 | 322 | 8862015 | 72 | |
BC138994.1 | - | 2 - 3 | 322 | 8862016 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |