Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172497.3 | NP_766085.2 | 6 - 7 | 1520 | 139 | |
GenBank mRNA | AK029550.1 | - | 6 - 7 | 1521 | 139 | |
AK167235.1 | - | 3 - 4 | 1148 | 139 | ||
BC132103.1 | - | 6 - 7 | 1511 | 40130380 | 139 | |
BC137880.1 | - | 6 - 7 | 1511 | 8860899 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |