Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172064.1 | NP_001165535.1 | 9 - 10 | 1539 | 80 | |
NM_001172065.1 | NP_001165536.1 | 9 - 10 | 1539 | 80 | ||
NM_153777.2 | NP_722472.1 | 9 - 10 | 1125 | 80 | ||
GenBank mRNA | AK017603.1 | - | 9 - 10 | 1513 | 80 | |
AK220237.1 | - | 9 - 10 | 1493 | 80 | ||
BC027807.1 | - | 9 - 10 | 1135 | 5324022 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |