Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181517.3 | NP_852658.2 | 21 - 22 | 2487 | 73 | |
GenBank mRNA | AK039534.1 | - | 21 - 22 | 2639 | 73 | |
AY316153.1 | - | 21 - 22 | 2487 | 73 | ||
BC053524.1 | - | 18 - 19 | 2170 | 73 | ||
BC064825.1 | - | 12 - 13 | 1443 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |