Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700037243.mm.1 |
NM_001286030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037243.mm.1 |
NM_001286031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037243.mm.1 |
NM_001286032.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037243.mm.1 |
NM_133941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037243.mm.1 |
NM_133941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037243.mm.1 |
TC0700004429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037253.mm.1 |
NM_001286032.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037253.mm.1 |
NM_133941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037253.mm.1 |
NM_133941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037253.mm.1 |
TC0700004429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037238.mm.1 |
NM_001286030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037238.mm.1 |
NM_001286031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037238.mm.1 |
NM_001286032.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037238.mm.1 |
NM_133941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037238.mm.1 |
NM_133941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037238.mm.1 |
TC0700004429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037241.mm.1 |
NM_001286030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037241.mm.1 |
NM_001286031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037241.mm.1 |
NM_001286032.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037241.mm.1 |
NM_133941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037241.mm.1 |
NM_133941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037241.mm.1 |
TC0700004429.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037255.mm.1 |
NM_001286032.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037255.mm.1 |
NM_133941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037255.mm.1 |
NM_133941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037255.mm.1 |
TC0700004429.mm.1 |
MTA TC |