Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019838.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019838.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019838.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019839.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019839.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019839.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019846.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019846.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019846.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019834.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019834.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019834.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019837.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019837.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019837.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019841.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019841.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019841.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019851.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019851.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019851.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019849.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019849.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019849.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019852.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019852.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019852.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019836.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019836.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019836.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019842.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019842.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019842.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019843.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019843.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019843.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019848.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019848.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019848.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019854.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019854.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019854.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019840.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019840.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019840.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019853.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019853.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019853.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019835.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019835.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019835.mm.1 |
TC1700002120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019844.mm.1 |
NM_009847.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019844.mm.1 |
NM_009847.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019844.mm.1 |
TC1700002120.mm.1 |
MTA TC |