Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048176.1 | NP_001041641.1 | 3 - 4 | 660 | 76 | |
GenBank mRNA | BC046474.1 | - | 3 - 4 | 766 | 76 | |
CJ130836.1 | - | 2 - 3 | 239 | 76 | ||
HQ426680.1 | - | 3 - 4 | 660 | 76 | ||
HQ426685.1 | - | 3 - 4 | 660 | 76 | ||
HQ426686.1 | - | 3 - 4 | 660 | 76 | ||
HQ426687.1 | - | 3 - 4 | 791 | 76 | ||
HQ426688.1 | - | 3 - 4 | 660 | 76 | ||
HQ426691.1 | - | 459 | 76 | |||
HQ426692.1 | - | 590 | 76 | |||
HQ426693.1 | - | 500 | 76 | |||
HQ426695.1 | - | 632 | 76 | |||
HQ426696.1 | - | 632 | 76 | |||
HQ426697.1 | - | 632 | 76 | |||
HQ426699.1 | - | 632 | 76 | |||
HQ426700.1 | - | 763 | 76 | |||
HQ426701.1 | - | 632 | 76 | |||
HQ426702.1 | - | 2 - 3 | 242 | 76 | ||
HQ426704.1 | - | 2 - 3 | 242 | 76 | ||
HQ426705.1 | - | 2 - 3 | 242 | 76 | ||
HQ426706.1 | - | 1 - 2 | 186 | 76 | ||
HQ426707.1 | - | 1 - 2 | 186 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |