Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048176.1 | NP_001041641.1 | 8 - 9 | 1184 | 105 | |
GenBank mRNA | BC046474.1 | - | 8 - 9 | 1290 | 105 | |
DT912627.1 | - | 4 - 5 | 380 | 105 | ||
HQ426680.1 | - | 8 - 9 | 1184 | 105 | ||
HQ426681.1 | - | 6 - 7 | 977 | 105 | ||
HQ426682.1 | - | 5 - 6 | 845 | 105 | ||
HQ426683.1 | - | 4 - 5 | 602 | 105 | ||
HQ426684.1 | - | 7 - 8 | 1052 | 105 | ||
HQ426686.1 | - | 7 - 8 | 1041 | 105 | ||
HQ426687.1 | - | 8 - 9 | 1315 | 105 | ||
HQ426688.1 | - | 8 - 9 | 1331 | 105 | ||
HQ426689.1 | - | 7 - 8 | 1199 | 105 | ||
HQ426690.1 | - | 8 - 9 | 1401 | 105 | ||
HQ426691.1 | - | 983 | 105 | |||
HQ426692.1 | - | 1114 | 105 | |||
HQ426693.1 | - | 1024 | 105 | |||
HQ426694.1 | - | 937 | 105 | |||
HQ426695.1 | - | 1156 | 105 | |||
HQ426697.1 | - | 1156 | 190 | |||
HQ426698.1 | - | 949 | 105 | |||
HQ426699.1 | - | 1013 | 105 | |||
HQ426700.1 | - | 1287 | 105 | |||
HQ426701.1 | - | 1303 | 105 | |||
HQ426702.1 | - | 7 - 8 | 766 | 105 | ||
HQ426703.1 | - | 5 - 6 | 559 | 105 | ||
HQ426704.1 | - | 7 - 8 | 766 | 105 | ||
HQ426705.1 | - | 6 - 7 | 623 | 105 | ||
HQ426706.1 | - | 6 - 7 | 710 | 105 | ||
HQ426707.1 | - | 6 - 7 | 857 | 105 | ||
HQ426709.1 | - | 4 - 5 | 458 | 105 | ||
HQ426711.1 | - | 4 - 5 | 605 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |