Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010811.2 | NP_034941.2 | 7 - 8 | 2309 | 82 | |
GenBank mRNA | AF074925.1 | - | 5 - 6 | 1559 | 82 | |
AK150100.1 | - | 7 - 8 | 2090 | 82 | ||
BC050829.1 | - | 7 - 8 | 2074 | 82 | ||
BC110480.1 | - | 5 - 6 | 1707 | 40048115 | 82 | |
U02304.1 | - | 6 - 7 | 2071 | 82 | ||
X75885.1 | - | 5 - 6 | 1713 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |