Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159548.1 | NP_001153020.1 | 3 - 4 | 415 | 77 | |
NM_001159549.1 | NP_001153021.1 | 3 - 4 | 415 | 77 | ||
NM_010264.4 | NP_034394.1 | 5 - 6 | 723 | 77 | ||
GenBank mRNA | AF390896.1 | - | 6 - 7 | 868 | 77 | |
CN537360.1 | - | 3 - 4 | 391 | 77 | ||
CN537407.1 | - | 3 - 4 | 415 | 77 | ||
CN538225.1 | - | 4 - 5 | 394 | 77 | ||
U09563.1 | - | 5 - 6 | 723 | 77 | ||
U14666.1 | - | 5 - 6 | 682 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mus musculus Probe spans exons |