Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172436.3 | NP_766024.1 | 6 - 7 | 645 | 79 | |
GenBank mRNA | AK019150.1 | - | 6 - 7 | 80 | ||
AK030595.1 | - | 6 - 7 | 675 | 79 | ||
AK083156.1 | - | 6 - 7 | 646 | 79 | ||
AK144958.1 | - | 6 - 7 | 679 | 79 | ||
AK147948.1 | - | 6 - 7 | 660 | 79 | ||
BC013845.1 | - | 6 - 7 | 624 | 79 | ||
BC024952.1 | - | 6 - 7 | 840 | 79 | ||
BC060505.1 | - | 6 - 7 | 632 | 6311836 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |