Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Api5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK134480.1 | - | 12 - 13 | 1482 | 73 | |
AK137466.1 | - | 12 - 13 | 1511 | 73 | ||
AK153934.1 | - | 12 - 13 | 1455 | 73 | ||
AK160874.1 | - | 12 - 13 | 1480 | 73 | ||
AK167315.1 | - | 12 - 13 | 1454 | 73 | ||
AK167375.1 | - | 12 - 13 | 1495 | 73 | ||
AK217064.1 | - | 3 - 4 | 184 | 73 | ||
BC007133.1 | - | 12 - 13 | 1474 | 3594358 | 73 | |
U35846.1 | - | 12 - 13 | 1439 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |