Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175113.3 | NP_780322.2 | 4 - 5 | 577 | 104 | |
GenBank mRNA | AK014359.1 | - | 4 - 5 | 574 | 104 | |
AK017672.1 | - | 4 - 5 | 519 | 104 | ||
AK028755.1 | - | 4 - 5 | 577 | 104 | ||
AK129300.1 | - | 4 - 5 | 544 | 104 | ||
AK164396.1 | - | 4 - 5 | 510 | 104 | ||
AK165976.1 | - | 4 - 5 | 529 | 104 | ||
AK167222.1 | - | 4 - 5 | 543 | 104 | ||
BC049083.1 | - | 1 - 2 | 215 | 104 | ||
BC052648.1 | - | 4 - 5 | 495 | 30030801 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |