Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101488.1 | NP_001094958.1 | 2 - 3 | 599 | 92 | |
GenBank mRNA | AK082731.1 | - | 2 - 3 | 601 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |