Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081117.2 | NP_001074586.2 | 11 - 12 | 2476 | 69 | |
GenBank mRNA | AK052772.1 | - | 11 - 12 | 2396 | 69 | |
AK082099.1 | - | 11 - 12 | 2395 | 69 | ||
X82786.1 | - | 11 - 12 | 2584 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |