Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011538.2 | NP_035668.2 | 4 - 5 | 795 | 65 | |
GenBank mRNA | AY654733.1 | - | 4 - 5 | 771 | 65 | |
BC140951.1 | - | 4 - 5 | 795 | 9055563 | 65 | |
U57331.1 | - | 4 - 5 | 795 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |