Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026992.3 | NP_081268.1 | 3 - 4 | 739 | 126 | |
GenBank mRNA | AK006521.1 | - | 3 - 4 | 740 | 126 | |
AY028460.1 | - | 3 - 4 | 421 | 126 | ||
BC030072.1 | - | 3 - 4 | 447 | 3376358 | 126 | |
BC091774.1 | - | 3 - 4 | 305 | 5150845 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |